Learn More
SPEX™ Silicon Nitride Ball
Product Code. 15370532 Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
8008 Silicon Nitride Grinding Vial Set
Unit Size:
Each
Product Code. 15370532
Brand: SPEX™ 8008A
This item requires a price update prior to being ordered.
Request a Price for this item here.
SPEX™ Silicon Nitride Ball is a replacement ball for use with 8008 Silicon Nitride Grinding Vial Set.
  • Extremely high resistance to abrasion and very long-wearing due to superior hardness
  • Moderate efficiency but very low levels of contamination
  • Typical composition: Major Elements – Si; Minor Elements – Y, Al, Fe, Ca
  • Hardness: Mohs: 8 1/2 + Knoop 1600
Specifications
Diameter (Metric) 12.7 mm
For Use With (Equipment) 8008 Silicon Nitride Grinding Vial Set
Material Silicon Nitride