Learn More
Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Description
Silicon Standard Metal Solution 1000ppm, for AAS, Solution Aqueous
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Color | Colorless |
Physical Form | Liquid |