Description
- Manufactured from FKM fluorocarbon
- Temperature-resistant up to 200°C
Specifications
Specifications
| Description | Door gasket - made of FKM, silicone-free |
| For Use With (Equipment) | BINDER Chambers ED 56 |
| Material | FKM Fluorocarbon |
| Temperature Range (Metric) | Ambient to 200°C |
| Type | Door Gasket |