Learn More
BINDER™ Door Gasket
Product Code. 15682046 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers ED 56
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15682046 BINDER Chambers ED 56 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15682046 Supplier BINDER™ Supplier No. 80121008
Material: FKM Fluorocarbon; For Use With: BINDER units ED 56
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Description Door gasket - made of FKM, silicone-free
For Use With (Equipment) BINDER Chambers ED 56
Material FKM Fluorocarbon
Temperature Range (Metric) Ambient to 200°C
Type Door Gasket