Learn More
Borer Chemie™ Silicone-free Grease Tube
Standard quality for desiccators, ground joints, stopcocks and apparatuses. Borer Chemie™ Silicone-Free Grease Tubes are suitable for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metal.
Brand:  Borer Chemie™ 514310.00-CA15
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 10343741
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Excellent thermal stability
  • Available in standard quality (vacuum tight up to 300°C) and high-vacuum quality (up to 400°C)
  • Constant lubricating consistency between -40°C and +250°C
  • No dripping point
  • Chemically stable against most acids, alkalis and gases
  • Insoluble in water and low molecular mono- and polyhydric alcohols
Specifications
Specifications
High-Vacuum
60 g