Learn More
Obermeier™ Silicone Low Viscosity Grease
Click to view available options
Quantity:
35 g
Unit Size:
35g
Specifications
Specifications
| Product Type | Low Viscosity Grease |
| Format | Paste |
| For Use With (Application) | Can be used as lubricants, water repellents, sealants and release agents |
| Certifications/Compliance | DIN 51376, DIN 51794 |
| Quantity | 35 g |
| Temperature (Metric) | -50°C to +200°C |
| Weight (Metric) | 35 g |