Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
Product Code. 10262161
Change view
Click to view available options
Diameter (Metric) Bottom:
18 mm
26 mm
41 mm
50.5 mm
56 mm
60 mm
64.5 mm
71 mm
79 mm
94 mm
Diameter (Metric) Top:
24 mm
32 mm
49 mm
59.5 mm
65 mm
70 mm
75.5 mm
83 mm
92 mm
107 mm
Unit Size:
Each
Pack of 100
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom Diameter (Metric) Top unitSize
10262161 71 mm 83 mm Each
10485071 26 mm 32 mm Pack of 100
10717781 18 mm 24 mm Each
10180221 41 mm 49 mm Each
10588321 56 mm 65 mm Each
10373951 64.5 mm 75.5 mm Each
10046401 79 mm 92 mm Each
10406321 94 mm 107 mm Each
10757971 50.5 mm 59.5 mm Each
10788351 60 mm 70 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10262161 Supplier Deutsch Neumann™ Supplier No. 1010571
Only null left
Add to basket
Add to basket
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Quantity 1/Ea.
Color Transparent
Diameter (Metric) Bottom 71 mm
Diameter (Metric) Top 83 mm
Material Silicone
Product Type Silicone Stopper