Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand: Deutsch Neumann™ 1010526
Additional Details : Weight : 2.19000kg
Description
- Acc. to DIN 12 871
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
Transparent | |
26 mm | |
1/Ea. | |
200°C |
Stopper | |
32 mm | |
Silicone |