Learn More
Hirschmann™ SILICOSEN™ Pink Silicone Stoppers
Product Code. 10238730
Change view
Click to view available options
Diameter (Metric) Inner:
6-9 mm
9-11 mm
11-14 mm
13-16 mm
15-18 mm
18-21 mm
20-23 mm
22-30 mm
30-40 mm
40-50 mm
Length (Metric):
10 mm
12 mm
13 mm
15 mm
20 mm
30 mm
35 mm
40 mm
Unit Size:
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) unitSize
10238730 13-16 mm 30 mm Pack of 10
10302843 6-9 mm 13 mm Pack of 10
10159722 11-14 mm 15 mm Pack of 10
10402861 13-16 mm 10 mm Pack of 10
10169722 15-18 mm 12 mm Pack of 10
10077842 18-21 mm 15 mm Pack of 10
10650644 20-23 mm 20 mm Pack of 10
10455421 22-30 mm 20 mm Pack of 10
10660644 30-40 mm 35 mm Pack of 10
10756263 40-50 mm 40 mm Pack of 10
10535683 9-11 mm 13 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10238730 Supplier Hirschmann™ Supplier No. 8905617
Only null left
Add to basket
Add to basket
Excellent for the cultivation of aerobic microorganisms. Hirschmann Instruments™ SILICOSEN™ Silicone Stoppers are specially processed culture plugs with continuous bubbles for preparation, filling and sterilization of culture media.
  • Uniform pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Diameter (Metric) Inner 13-16 mm
For Use With (Application) For The Preparation And Sterilization Of Culture Media
Length (Metric) 30 mm
Quantity 10/Pk.
Color Pink
Material Silicone
Product Type Tapered Stopper