Learn More
B Braun™ Silkam™ Needle-Thread Combination
Product Code. 15317425
Change view
Click to view available options
Length (Metric):
0.7 mm
Needle Point Number:
DS 16
Shape:
3/8 Circle Reverse Cutting Needle
Size:
USP metric 6/0
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Needle Point Number Shape Size Unit Size
15317425 0.7 mm DS 16 3/8 Circle Reverse Cutting Needle USP metric 6/0 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15317425 Supplier B Braun™ Supplier No. C0762113
  • ONLINE EXCLUSIVE PRICE
    € 279.00
    € 276.19 / Pack of 1
    Save € 2.81
    1% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A natural, non-absorbable braided and coated suture made of natural silk filaments.
  • Indicated for applications where it is surgical practice to employ non-absorbable material
  • Excellent handling properties
  • Good knot security
  • Indications include general surgery, oral surgery, neurosurgery, skin closure and ligatures
Specifications
Material Silk
Product Type Needle-thread Suture Combo
Length (Metric) 0.7 mm
Size USP metric 6/0
For Use With (Application) Surgical
Color Black
Shape 3/8 Circle Reverse Cutting Needle
Needle Point Number DS 16