missing translation for 'onlineSavingsMsg'
Learn More

Silmitasertib, MedChemExpress

Product Code. 16474027
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16474027

Brand: Medchem Express HY50855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.

Specifications

Chemical Name or Material Silmitasertib
CAS 1009820-21-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H12ClN3O2
Quantity 50 mg
Synonym CX-4945
Solubility Information DMSO : ≥ 35 mg/mL (100.07 mM) ∣0.1 M NaOH : 33.33 mg/mL (95.29 mM; ultrasonic and adjust pH to 9 with NaOH)
SMILES O=C(C1=CC=C2C3=C(C(NC4=CC=CC(Cl)=C4)=NC2=C1)C=CN=C3)O
Molecular Weight (g/mol) 349.77
Formula Weight 349.77
Percent Purity 96.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.