Learn More
Silmitasertib, MedChemExpress
Product Code. 16480568
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16480568
Brand: Medchem Express HY50855
MedChemExpress Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.
Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'.
Specifications
Chemical Name or Material Silmitasertib
CAS 1009820-21-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H12ClN3O2
Quantity 5 mg
Synonym CX-4945
Solubility Information DMSO : ≥ 35 mg/mL (100.07 mM) ∣0.1 M NaOH : 33.33 mg/mL (95.29 mM; ultrasonic and adjust pH to 9 with NaOH)
SMILES O=C(C1=CC=C2C3=C(C(NC4=CC=CC(Cl)=C4)=NC2=C1)C=CN=C3)O
Molecular Weight (g/mol) 349.77
Formula Weight 349.77
Percent Purity 96.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only