missing translation for 'onlineSavingsMsg'
Learn More

Silodosin b-D-Glucuronide Sodium Salt (>90%), TRC

Product Code. 30426132
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426132 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426132 Supplier Toronto Research Chemicals Supplier No. TRCS4650105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silodosin beta-D-Glucuronide Sodium Salt
CAS 879292-24-7
Molecular Formula C31H39F3N3NaO10
Quantity 5 mg
Synonym 3-[7-(Aminocarbonyl)-2,3-dihydro-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indol-1-yl]propyl β-D-Glucopyranosiduronic Acid Monosodium Salt
InChI Formula InChI=1S/C31H40F3N3O10.Na/c1-17(36-8-12-44-21-5-2-3-6-22(21)46-16-31(32,33)34)13-18-14-19-7-10-37(23(19)20(15-18)28(35)41)9-4-11-45-30-26(40)24(38)25(39)27(47-30)29(42)43;/h2-3,5-6,14-15,17,24-27,30,36,38-40H,4,7-13,16H2,1H3,(H2,35,41)(H,42,43);/q;+1/p-1/t17-,24+,25+,26-,27+,30-;/m1./s1
SMILES C[C@H](CC1=CC2=C(C(=C1)C(=O)N)N(CC2)CCCO[C@H]3[C@@H]([C@H]([C@@H]([C@H](O3)C(=O)[O-])O)O)O)NCCOC4=CC=CC=C4OCC(F)(F)F.[Na+]
IUPAC Name sodium;(2S,3S,4S,5R,6R)-6-[3-[7-carbamoyl-5-[(2R)-2-[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethylamino]propyl]-2,3-dihydroindol-1-yl]propoxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol) 693.64
Formula Weight 693.2485
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.