Learn More
Buerkle™ Silo Sampler SiloPicker, Container
Buerkle™ Stainless Steel SiloPicker Sample Container is a spare part for Buerkle™ Stainless Steel SiloPicker Sampler.
Brand: Buerkle™ 5340-0100
352.84 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.89500kg
Description
- Stainless steel AISI 316L
- Spare container
Specifications
Sample Container | |
Autoclavable | |
100 mL |
Stainless Steel (V4A) | |
Equipment for Silo sampler SiloPicker | |
Silo Sampler SiloPicker |