Learn More
Trajan Scientific and Medical™ SilTite™ Column Connector Kit
Ensures easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073203
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248913
  • € 133.00 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
1/16 in. column
SilTite Kit 1/16 in. Column for PerkinElmer
10 ferrules, 2 nuts
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers