Learn More
Trajan Scientific and Medical™ SilTite™ Ferrules
Product Code. 15228923
Click to view available options
For Use With (Equipment):
Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns
Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns
Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns
Thermo GC/MS Interface Connections 0.32 mm I.D. Columns
Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns
Unit Size:
Pack of 10
Product Code. 15228923
Brand: Trajan Scientific and Medical™ 073229
Only null left
Add to basket
Add to basket
Ensures easy, leak-free installation for capillary columns without the use of any tools.
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type SilTite Ferrules for 0.53 mm ID Columns for Shimadzu
For Use With (Equipment) Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns