Learn More
Thermo Scientific™ SilTite™ GC Capillary Column Connector
Product Code. 10497514 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Type:
Capillary Column Connector
Replacement Column Connector Nuts
Diameter (Metric) Inner:
0.1 to 0.25 mm
0.1 to 0.53 mm
0.45 to 0.53 mm
32 mm
missing translation for 'unitSize'
Each
Pack of 5
Product Code. 10497514
missing translation for 'mfr': Thermo Scientific™ 290MU498
Only null left
Add to basket
Add to basket
Use SilTite GC Capillary Column Connectors for fused silica capillary GC columns.
Use Thermo Scientific™ SilTite Capillary Column Connectors for fused silica capillary GC columns. They are glass lined for inertness and have a low thermal mass.
Join together two GC columns
Repair broken columns
TRUSTED_SUSTAINABILITY
Specifications
Type Capillary Column Connector
Description 290 MU498
Diameter (Metric) Inner 0.1 to 0.25 mm
For Use With (Equipment) Fused Silica Capillary Column
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.