Learn More
Descripción
- The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
- Individually packed
- Grey Color
- With exhalation valve
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Soft sealing lip in the nose and chin area guarantees a secure, comfortable sealed position of the mask
- Adjustable head band offering easy handling and comfortable fit
- Flexible and adjustable nose clip
- Soft material edges for high wearing comfort Certified FFP3 face mask to EN 149 D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Additional activated carbon filter against odours, gases and vapours below threshold
- Cannot be reused
- Respirators are not suitable for people with beards, sideburns or deep scars near the seals
Especificaciones
Especificaciones
| Color | Gray |
| Design Type | Flat-fold |
| Disposable | Single-use |
| Filter Type | FFP3 |
| Material | Polyester |
| Product Type | Particulate Respirator |
| Size | Medium |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?