Learn More
Uvex Silv-Air Classic 2220 FFP2 Mask
Shop All Uvex Products
Click to view available options
:
Filter Type: FFP2
Description
- With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
- The masks can also be worn comfortably for a longer period of time without becoming bothersome.
Specifications
Specifications
| Aerosol Type | P - Oil-Proof |
| Color | Gray |
| Design Type | Cup/Molded |
| Disposable | Reusable |
| Filter Type | FFP2 |
| Material | Polypropylene, Polyester |
| Product Type | Healthcare Particulate Respirator and Surgical Mask |
| Size | Medium |
| With Exhalation Valve | Yes |
| Latex Free | Yes |
| Show More |