missing translation for 'onlineSavingsMsg'
Learn More

Silver carbonate, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-221891
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
534-16-7
CAg2O3
275.75
MFCD00003403
WIKQEUJFZPCFNJ-UHFFFAOYSA-N
carbonic acid; silver, carbonic acid disilver
25203940
carbonic acid;silver
C(=O)(O)O.[Ag].[Ag]
534-16-7
CAg2O3
275.75
MFCD00003403
WIKQEUJFZPCFNJ-UHFFFAOYSA-N
carbonic acid; silver, carbonic acid disilver
25203940
carbonic acid;silver
C(=O)(O)O.[Ag].[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 210.0°C
Color Green to Green-Yellow
Physical Form Granular Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula Ag2CO3
Fieser 01,1005; 02,363; 04,425; 06,511; 08,411; 12,432
Merck Index 15, 8646
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonium hydroxide and nitric acid, insoluble in alcohol
Formula Weight 275.75
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Silver carbonate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver carbonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'