missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540693
This item is not returnable.
View return policy
Description
Peptide sequence: SSSKSKSRTS PYPQYSGFHT ERSESDHDSQ WGGSPLTDTA SPQLLDPADR Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
SIM1 and SIM2 genes are Drosophila single-minded (sim) gene homologs. SIM1 transcript was detected only in fetal kidney out of various adult and fetal tissues tested. Since the sim gene plays an important role in Drosophila development and has peak levels of expression during the period of neurogenesis, it was proposed that the human SIM gene is a candidate for involvement in certain dysmorphic features (particularly the facial and skull characteristics), abnormalities of brain development, and/or mental retardation of Down syndrome.
Specifications
| SIM1 | |
| Polyclonal | |
| Unconjugated | |
| SIM1 | |
| BHLHE14; bHLH-PAS transcription factor; Class E basic helix-loop-helix protein 14; mSIM1; SIM bHLH transcription factor 1; SIM bHLH transcription factor 1a; sim1; sim1a; single-minded 1; single-minded family bHLH transcription factor 1; single-minded family bHLH transcription factor 1a; single-minded homolog 1; single-minded homolog 1 (Drosophila); single-minded homolog 1-A; single-minded-like 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20464 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q61045 | |
| SIM1 | |
| Synthetic peptide directed towards the middle region of human SIM1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction