missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simazine-d10 (diethyl-d10), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Simazine D10 |
| CAS | 220621-39-6 |
| Molecular Formula | C7 2H10 H2 Cl N5 |
| Quantity | 0.01 g |
| Synonym | Simazine D10 |
| InChI Formula | InChI=1 S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])Nc1nc(Cl)nc(NC([2 H])([2 H])C([2 H])([2 H])[2 H])n1 |
| IUPAC Name | 6-chloro-2-N,4-N-bis(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 211.72 |
| Formula Weight | 211.1409 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction