missing translation for 'onlineSavingsMsg'
Learn More

Simeprevir, MedChemExpress

Product Code. 16470117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470117 50 mg 50mg
16436467 5 mg 5mg
16424806 100 mg 100mg
16413738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470117 Supplier Medchem Express Supplier No. HY10241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Simeprevir (TMC435; TMC435350) is an oral, potent and highly specific hepatitis C virus (HCV) NS3/4A protease inhibitor with a Ki of 0.36 nM.

Simeprevir inhibits HCV replication with an EC50 of 7.8 nM. Simeprevir also potently suppresses SARS-CoV-2 replication and synergizes with Remdesivir. Simeprevir inhibits the main protease (Mpro) and the RNA-dependent RNA polymerase (RdRp) of SARS-CoV-2, and also modulates host immune responses.

Specifications

Chemical Name or Material Simeprevir
Physical Form Solid
CAS 923604-59-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C38H47N5O7S2
For Use With (Application) COVID-19-anti-virus
Synonym TMC435 TMC435350
Solubility Information DMSO : 14.29 mg/mL (19.05 mM; Need ultrasonic)
SMILES COC1=C(C)C2=C(C(O[C@H]3C[C@@H](C(N(C)CCCC/C=C\[C@H](C4)[C@]4(C(NS(=O)(C5CC5)=O)=O)N6)=O)[C@H](C6=O)C3)=CC(C7=NC(C(C)C)=CS7)=N2)C=C1
Molecular Weight (g/mol) 749.94
Formula Weight 749.94
Percent Purity 99.46%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.