Learn More
Motic™ Simple Add-on Polarizer Filter
Description
- For use with BA Series Microscopes
- Requires use with Analyzer
- Illuminates the specimen while analyzer restricts detected light
Specifications
Specifications
| Type | Polarizing Filter |
| Description | Requires use with Analyzer Filter |
| For Use With (Application) | Specimen illumination |
| For Use With (Equipment) | BA Series Microscopes |
| Quantity | 1 Ea. |
Product Title
Haben Sie Verbesserungsvorschläge?