missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin Nombre Virus Glycoprotein 2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41256UV
This item is not returnable.
View return policy
Description
Sin Nombre Virus Glycoprotein 2 Polyclonal antibody specifically detects Sin Nombre Virus Glycoprotein 2 in Virus samples. It is validated for ELISA
Specifications
| Sin Nombre Virus Glycoprotein 2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| DyLight 350 | |
| Envelope polyprotein, GPC, M polyprotein, SNVsMgp1 | |
| Antibody was raised against a 16 amino acid peptide near the center of the Sin Nombre virus glycoprotein. The immunogen is located within amino acids 670 - 720 of Sin Nombre Virus Glycoprotein 2. Amino Acid Squence: YSYRRKLVNPANQEET | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 2654026 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction