missing translation for 'onlineSavingsMsg'
Learn More

Sin Nombre Virus Nucleocapsid Antibody - BSA Free, Novus Biologicals™

Product Code. p-200075512 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18601159 0.025 mg 0.03mg
18145048 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18601159 Supplier Novus Biologicals Supplier No. NBP2412570.025mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sin Nombre Virus Nucleocapsid Polyclonal specifically detects Sin Nombre Virus Nucleocapsid in Virus samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sin Nombre Virus Nucleocapsid
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias Nucleoprotein, SNVsSgp1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the center of the Sin Nombre virus nucleocapsid. The immunogen is located within amino acids 170 - 220 of Sin Nombre Virus Nucleocapsid. Amino Acid Squence: PTAQSTMKADEITPGRFRT
Molecular Weight of Antigen 48 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.