missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sin Nombre Virus Nucleocapsid Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41257DL594
This item is not returnable.
View return policy
Description
Sin Nombre Virus Nucleocapsid Polyclonal antibody specifically detects Sin Nombre Virus Nucleocapsid in Virus samples. It is validated for ELISA
Specifications
| Sin Nombre Virus Nucleocapsid | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| DyLight 594 | |
| Nucleoprotein, SNVsSgp1 | |
| Antibody was raised against a 19 amino acid peptide near the center of the Sin Nombre virus nucleocapsid. The immunogen is located within amino acids 170 - 220 of Sin Nombre Virus Nucleocapsid. Amino Acid Squence: PTAQSTMKADEITPGRFRT | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 2943142 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction