missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIN3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98964-50ul
This item is not returnable.
View return policy
Description
SIN3A Polyclonal antibody specifically detects SIN3A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SIN3A | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human SIN3A. | |
| 50 μg | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience | |
| 25942 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction