Learn More
Sinefungin, TRC
Specifications
Specifications
| Chemical Name or Material | Sinefungin |
| CAS | 58944-73-3 |
| Molecular Formula | C15 H23 N7 O5 |
| Quantity | 1 mg |
| Synonym | 6,9-Diamino-1-(6-amino-9H-purin-9-yl)-1,5,6,7,8,9-hexadeoxy-D-glycero-α-L-talo-decofuranuronic Acid, 32232RP, A 9145, Antibiotic 32232RP, Antibiotic A 9145, Compound 57926, RP 32232 |
| InChI Formula | InChI=1S/C15H23N7O5/c16-6(1-2-7(17)15(25)26)3-8-10(23)11(24)14(27-8)22-5-21-9-12(18)19-4-20-13(9)22/h4-8,10-11,14,23-24H,1-3,16-17H2,(H,25,26)(H2,18,19,20)/t6-,7-,8+,10+,11+,14+/m0/s1 |
| SMILES | N[C@@H](CC[C@H](N)C(=O)O)C[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23 |
| IUPAC Name | (2S,5S)-2,5-diamino-6-[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]hexanoic acid |
| Molecular Weight (g/mol) | 381.387 |
| Formula Weight | 381.176 |