Learn More
Buerkle™ Single Mark Plastic Pipets
Product Code. 11911531 Shop All Bürkle Products
Click to view available options
Volume (Metric):
50 mL
Color:
Transparent
Unit Size:
Each
Product Code. 11911531
Brand: Buerkle™ 75530050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry
  • PP, transparent
  • Blue, permanently coloured graduation and labelling
TRUSTED_SUSTAINABILITY
Specifications
Class Pipette
Material Polypropylene
For Use With (Application) For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used, e.g. in Fluorine and Silicate Chemistry
Graduated Yes
Quantity 1
Color Transparent
Volume (Metric) 50 mL
Type One-mark Pipet