Learn More
Buerkle™ Single Mark Plastic Pipets
Shop All Bürkle Products
Click to view available options
Volume (Metric):
50 mL
Color:
Transparent
Unit Size:
Each
Description
- PP, transparent
- Blue, permanently coloured graduation and labelling
Specifications
Specifications
| Class | Pipette |
| Material | Polypropylene |
| For Use With (Application) | For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used, e.g. in Fluorine and Silicate Chemistry |
| Graduated | Yes |
| Quantity | 1 |
| Color | Transparent |
| Volume (Metric) | 50 mL |
| Type | One-mark Pipet |