Learn More
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.060g/cm3
Product Code. 10313902
Click to view available options
Specific Gravity Range:
0.940 to 1.000 g/cm3
1.060 to 1.120 g/cm3
1.120 to 1.180 g/cm3
1.180 to 1.240 g/cm3
1.300 to 1.360 g/cm3
1.360 to 1.420 g/cm3
1.480 to 1.540 g/cm3
Unit Size:
Each
Product Code. 10313902
Brand: Amarell™ H800016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Calibrated for reference temperature 20°C. The standard density hydrometer without thermometer can be used for many general specific gravity measurements.
  • Reference temperature 20°C
  • Each hydrometer length is 160mm
Specifications
Quantity 1Each
Type Density Hydrometer
Accuracy ±1g/cm3
Increments 0.001g
Length (Metric) 160 mm
Specific Gravity Range 1.060 to 1.120 g/cm3
Temperature of Standardization 20°C