Learn More
Amarell™ Standard Density Hydrometer without Thermometer, measuring span 0.060g/cm3
Calibrated for reference temperature 20°C. The standard density hydrometer without thermometer can be used for many general specific gravity measurements.
Brand:  Amarell™ H800018
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 10373942
  • € 44.45 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Reference temperature 20°C
  • Each hydrometer length is 160mm
Specifications
Specifications
1Each
±1g/cm3
160 mm
20°C
Density Hydrometer
0.001g
1.120 to 1.180 g/cm3