Learn More
SINGLET™ Single-Use pH Buffer, pH 10.01 (NIST), Hach Lange™
Click to view available options
Quantity:
20 Pk.
Unit Size:
Each
Description
Hach Lange SINGLET Single-Use pH Buffer, pH 10.01 (NIST), Quantity: 20 pk., Blue, pH: 10.01
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Color | Blue |
| pH | 10.01 |
| Physical Form | Liquid |
| Quantity | 20 Pk. |
| Solution Type | Buffer |
| Formulation | Water, Sodium Carbonate, Methyl Alcohol, Formaldehyde, Cuprate (2-), [29h, 31h-Phthalocyanine-C, C-Disulfonato (4-)-N29, N30, N31, N32], Disodium, Sodium Bicarbonate |
| Solubility Information | Soluble |