missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SINGLET™ Single-Use pH Buffer, pH 4.01 (NIST), Hach Lange™
Click to view available options
Quantity:
20 Pk.
Unit Size:
Each
Description
Hach Lange SINGLET Single-Use pH Buffer, pH 4.01 (NIST), Quantity: 20 pk., Red, pH: 4.01
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Red |
| pH | 4.01 |
| Physical Form | Liquid |
| Quantity | 20 Pk. |
| Solution Type | Buffer |
| Formulation | Water, Monopotassium Salt, Amaranth Red No. 27, Benzethonium Chloride, 1,2-Benzenedicarboxylic Acid |
| Solubility Information | Soluble in Water |
Safety and Handling
Product Identifier
- Singlet pH Buffer Solution; pH 4.01
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction