missing translation for 'onlineSavingsMsg'
Learn More

ARG1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human ARG1 (P05089-1) (Met 1-Lys 322) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11558H08H25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15248655

  • € 530.00 / 25µg


Estimated Shipment
21-08-2020
Add to basket

Description

Description

Arginase Liver (ARG1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by the production of urea during the hydrolysis of arginine; the specific activity is >35,000pmole/min./μg
  • Formulation: 20mM Tris, 150mM NaCl, 0.2μg/μL BSA, 20% glycerol, 1mM DTT, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Assay Standard
Frozen
≥85% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ARG1
ARG1
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
36.2kDa
5 x 5μg
<1 EU/μg
Arginase, liver
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.