missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ CES2E (CES5) Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CES2E (NP_766347.1) (Met 1-His 556) fused to the polyhistidine tag at the C-terminus

Merk:  Thermo Scientific™ 50620M08H250

Overige gegevens : Gewicht : 0.10000kg

 Bekijk meerdere types van dit product via de

Produktcode. 15359046

  • € 3210.00 / 250 µg


Geschatte verzenddatum
bij leverancier 25-09-2018
Toevoegen aan winkelmandje

Inclusief: Five vials of lyophilized protein

Beschrijving

Beschrijving

Carboxylesterase 2E (CES2E), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to hydrolyze pnitrophenylacetate; the specific activity is >50nM/min./μg
  • Formulation: Lyophilized in 20mM Tris,150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specificatie

Specificatie

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
60.6kDa
5 x 50μg
Other Proteins
9030624L02Rik, CES5
CES2E (CES5)
Other Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Carboxylesterase 2E
Assay Standard
Mammalian
Video's

For Research Use Only. Not for use in diagnostic procedures.