missing translation for 'onlineSavingsMsg'
Learn More

USP7 Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the amino acid sequence (Lys 208-Glu 560) of human USP7 (NP_003461.2) fused to two additional aa (Gly & Pro) at the N-terminus

Brand:  Invitrogen 11681HNCB250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15248835

  • € 190.00 / 250µg


Estimated Shipment
13-08-2020
Add to basket

Description

Description

Ubiquitin Carboxyl-Terminal Hydrolase 7 (USP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Insect
Assay Standard
Lyophilized
41kDa
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Ubiquitin carboxyl-terminal hydrolase 7
Proteases, Peptidases & Inhibitors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
7874
PAGE
5 x 50μg
Truncated/Recombinant
HAUSP, TEF1
USP7
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.