Learn More
Invitrogen™ Sintilimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559732
 View more versions of this product
Product Code. 30283223
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sintilimab is a fully human anti-PD-1 antibody for tumor immunotherapy. It adopts IgG4 molecular framework and modified its hinge region sequence (S228P). Sintilimab, as a programmed death receptor 1 (PD-1) blocker, can bind to PD-1 molecules on the surface of T cells and relieve the inhibitory effect of PD-1 pathway on T cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sintilimab
Recombinant Monoclonal
Unconjugated
IBI-308
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized