Learn More
Invitrogen™ SIPA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588447
Description
Immunogen sequence: ESGRPRRSFS ELYTLSLQEP SRRGAPDPVQ DEVQGVTLLP TTKQLLHLCL QDGGSPPGPG DLAEERTEFL HSQNSLSPRS SLSDEAPVLP NTTPDLLLAT TAKPSVPSAD SETPLTQDRP GSPSGSEDKG NPAPELRASF LPRTLSLRNS ISRIMSEAGS GTLEDEWQAI SEIASTCNTI LESLSREGQP IPESGDPKGT PKSDAEPEPG NLSEKVSHLE SMLRKLQEDL QKEKADRAAL EEEVRSLRHN NRRLQAESES AATRLLLASK QLGSPTADLA; Positive Samples: Raji, K-562, HeLa, LO2, Mouse spleen; Cellular Location: Cytoplasm, Endomembrane system, Nucleus, Peripheral membrane protein, perinuclear region.
The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.
Specifications
| SIPA1 | |
| Polyclonal | |
| Unconjugated | |
| SIPA1 | |
| GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20469, 6494 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P46062, Q96FS4 | |
| SIPA1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 763-1042 of human SIPA1 (NP_69498529). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.