missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIPA1L1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81801AF750
This item is not returnable.
View return policy
Description
SIPA1L1 Polyclonal antibody specifically detects SIPA1L1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SIPA1L1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| E6-targeted protein 1, FLJ37213, H_DJ1140G11.1, high-risk human papilloma viruses E6 oncoproteins targeted protein 1, KIAA0440, signal-induced proliferation-associated 1 like 1 | |
| SIPA1L1 antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SIPA1L1. The immunogen is located within amino acids 40 - 90 of SIPA1L1. Amino Acid Squence: PVGPPRSEGSHHITS | |
| 0.1 mL | |
| Cancer | |
| 26037 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction