Learn More
SIPr (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | SIPr (Technical Grade) |
| CAS | 258278-28-3 |
| Molecular Formula | C27H38N2 |
| Quantity | 500 mg |
| Synonym | 1,3-Bis[2,6-bis(1-methylethyl)phenyl]-2-imidazolidinylidene, 1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydroimidazol-2-ylidene, 1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene |
| InChI Formula | InChI=1S/C27H38N2/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8/h9-14,18-21H,15-16H2,1-8H3 |
| SMILES | CC(C1=C(N2[C]N(C3=C(C(C)C)C=CC=C3C(C)C)CC2)C(C(C)C)=CC=C1)C |
| IUPAC Name | 1,3-bis(2,6-diisopropylphenyl)-4,5-dihydro-1H-imidazol-3-ium-2-ide |
| Molecular Weight (g/mol) | 390.6 |
| Formula Weight | 390.3 |