missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SIRP alpha/CD172a Antibody (BI 765063) - Humanized, IgG4PE, Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 920.00
Specifications
| Antigen | SIRP alpha/CD172a |
|---|---|
| Clone | BI 765063 |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047469
|
Novus Biologicals™
NBP3-28462-100ug |
100 μg |
€ 337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30044545
|
Novus Biologicals™
NBP3-28462-50ug |
50 μg |
€ 213.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30046398
|
Novus Biologicals™
NBP3-28462-1mg |
1 mg |
€ 920.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIRP alpha/CD172a Monoclonal antibody specifically detects SIRP alpha/CD172a in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| SIRP alpha/CD172a | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| GPCR, Protein Phosphatase, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 | |
| SIRPa/ CD172a | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| BI 765063 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P78324 | |
| 140885 | |
| IgG4 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title