missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP alpha/CD172a Antibody (OTI7B3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74197B
This item is not returnable.
View return policy
Description
SIRP alpha/CD172a Monoclonal specifically detects SIRP alpha/CD172a in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SIRP alpha/CD172a | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| SIRPA | |
| Full length recombinant protein of human SIRP alpha/CD172a (NP_001035111) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI7B3 | |
| Western Blot, Immunohistochemistry | |
| Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 | |
| Mouse | |
| Affinity Purified | |
| GPCR, Protein Phosphatase, Signal Transduction | |
| 140885.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction