Learn More
SIRP beta 1/CD172b Antibody (308908), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB20961E
Product Code. 18520635
  • ONLINE EXCLUSIVE PRICE
    € 618.00
    € 583.80 / 0.10mL
    Save € 34.20
    5% Off
Description
SIRP beta 1/CD172b Monoclonal antibody specifically detects SIRP beta 1/CD172b in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SIRP beta 1/CD172b
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, Immunocytochemistry, CyTOF
308908
Flow Cytometry, Immunocytochemistry, CyTOF-ready
CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRP-BETA-1, SIRP-beta-1 isoform 3
Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4
0.1 mL
Signal Transduction
10326
Store at 4°C in the dark.