missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP beta 1/CD172b Antibody (308908), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB20961P
This item is not returnable.
View return policy
Description
SIRP beta 1/CD172b Monoclonal antibody specifically detects SIRP beta 1/CD172b in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| SIRP beta 1/CD172b | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 308908 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
| Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4 | |
| 0.1 mL | |
| Signal Transduction | |
| 10326 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction