missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP beta 1/gamma Antibody (308906), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2096MFV450
This item is not returnable.
View return policy
Description
SIRP beta 1/gamma Monoclonal antibody specifically detects SIRP beta 1/gamma in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| SIRP beta 1/gamma | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cellular Signaling, Immunology | |
| Human | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 308906 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD172b;SIRPB1;SIRP-BETA-1 | |
| Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4 | |
| RUO | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction