missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP gamma/CD172g Antibody (1H7C2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06476UV
This item is not returnable.
View return policy
Description
SIRP gamma/CD172g Monoclonal antibody specifically detects SIRP gamma/CD172g in Human samples. It is validated for ELISA
Specifications
| SIRP gamma/CD172g | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1H7C2 | |
| ELISA | |
| bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIRP gamma/CD172g (NP_061026.2; Met 1-Ser 364). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 55423 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction