Learn More
Novus Biologicals™ SIRP gamma/CD172g Antibody (KWAR23) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28671-100ug
 View more versions of this product
Product Code. 30047217
  • € 338.00 / 100µg
Description
SIRP gamma/CD172g Monoclonal antibody specifically detects SIRP gamma/CD172g in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SIRP gamma/CD172g
Monoclonal
Unconjugated
Q9P1W8
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG4
Flow Cytometry, ELISA, Functional Assay
KWAR23
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma
SIRPg/ CD172g
100 μg
Cell Cycle and Replication
55423