missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRP gamma Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 156.00 - € 453.00
Specifications
| Antigen | SIRP gamma |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17773004
|
Invitrogen™
PA5121609 |
20 μL |
€ 156.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17763004
![]() |
Invitrogen™
PA5121608 |
100 μL |
€ 453.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SIRP gamma (CD172g) recognizes a 55 kDa signal regulatory protein which is a member of the immunoglobulin gene superfamily. SIRP gamma is expressed on most T lymphocytes and a subset of B-cells but is absent on myeloid cells. CD172g has a truncated cytoplasmic tail, which is similar to SIRP beta, but unlike SIRP beta CD172g does not require DAP12 for expression at the cell surface. CD172g, like SIRP alpha, binds to CD47 inducing apoptosis. SIRP gamma is also involved in the negative regulation of receptor tyrosine kinase-coupled signalling.Specifications
| SIRP gamma | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9P1W8 | |
| 55423 | |
| Pro111-Ile372 (Accession Q9P1W8), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SIRPG |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| bA77C3.1; CD172 antigen-like family member B; CD172 gamma; CD172g; CD172g antigen; RP11-77C3.2; signal regulatory protein gamma; signal-regulatory protein beta 2; signal-regulatory protein beta-2; Signal-regulatory protein gamma; SIRP beta 2; SIRPB2; SIRP-B2; SIRP-beta-2; SIRPG; SIRPgamma; SIRP-gamma | |
| SIRPG | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title