missing translation for 'onlineSavingsMsg'
Learn More

SIRPA Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene

Product Code. p-200015049
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17587212 30 μL 30µL
17567212 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17587212 Supplier Origene Technologies Supplier No. TA507333S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SIRPA Monoclonal antibody specifically detects SIRPA in Human samples. It is validated for Western Blot

Specifications

Antigen SIRPA
Applications Western Blot
Classification Monoclonal
Clone OTI1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ankmy2
Gene Accession No. Q8IV38
Gene Alias AI835480; Bit; brain Ig-like molecule with tyrosine-based activation motifs; Brain immunoglobulin like protein with tyrosine - based activation motifs; brain immunological-like with tyrosine-based motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172; CD172 alpha; CD172 antigen-like family member A; CD172A; Inhibitory receptor SHPS-1; macrophage fusion receptor; Macrophage membrane protein MFP150; MFR; mSIRP-alpha1; Myd1; MYD-1; myD-1 antigen; p84; Protein tyrosine phosphatase non-receptor type substrate 1 (SHP substrate 1); protein tyrosine phosphatase, non-receptor type substrate 1; Protein tyrosine phosphatase, non-receptor type substrate 1 (SHP substrate 1); Ptpns1; SHP substrate 1; SHP-1; SHPS1; SHPS-1; signal regulatory protein alpha; signal-regulatory protein alpha; signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; SIRP; Sirpa; SIRPalpha; Sirp-alpha-1; SIRPalpha2; Sirp-alpha-2; Sirp-alpha-3; swc3; swine workshop cluster 3 antigen; swine workshop cluster 3 antigen precursor; tyrosine phosphatase SHP substrate 1; tyrosine-protein phosphatase non-receptor type substrate 1
Host Species Mouse
Immunogen Full length human recombinant protein of human ANKMY2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57037
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.